missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€ - 619.00€
Specifications
| Antigen | Transferrin |
|---|---|
| Dilution | Western Blot 1:2000, Immunoprecipitation 1:10-1:500 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067948
|
Novus Biologicals
NBP1-97472-0.025MG |
0.025 mg |
204.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18102830
|
Novus Biologicals
NBP1-97472-0.05MG |
0.05 mg |
418.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18018367
|
Novus Biologicals
NBP1-97472-0.2MG |
0.2 mg |
619.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transferrin Polyclonal specifically detects Transferrin in Human, Mouse, Rat, Canine, Guinea Pig samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Transferrin | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Canine, Rat, Guinea Pig | |
| P02787 | |
| 7018 | |
| Human Transferrin. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:2000, Immunoprecipitation 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin | |
| TF | |
| IgG | |
| Protein A purified | |
| 77 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title