missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Transferrin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-97472-0.025MG
This item is not returnable.
View return policy
Description
Transferrin Polyclonal specifically detects Transferrin in Human, Mouse, Rat, Canine, Guinea Pig samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Transferrin | |
| Polyclonal | |
| Western Blot 1:2000, Immunoprecipitation 1:10-1:500 | |
| P02787 | |
| TF | |
| Human Transferrin. | |
| Protein A purified | |
| RUO | |
| 7018 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin | |
| Rabbit | |
| 77 kDa | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Canine, Rat, Guinea Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction