missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenyl acetate, 98%

Product Code. 11453068
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11453068

Brand: Thermo Scientific Alfa Aesar A18786.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2497-18-9 | C8H14O2 | 142.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2497-18-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00009474
InChI Key HRHOWZHRCRZVCU-AATRIKPKSA-N
Synonym z-hex-2-enyl acetate, unii-pt792bq57d, 2-hexen-1-ol, acetate, z, z-2-hexenyl acetate, fema no. 2564, 2-hexen-1-yl acetate, trans, 2-hexen-1-ol, acetate, 2e, 2-hexenyl acetate, trans, hexenyl acetate, 2z
PubChem CID 2733294
IUPAC Name [(E)-hex-2-enyl] acetate
SMILES CCCC=CCOC(=O)C

Specifications

Density 0.9
Boiling Point 165°C to 166°C
Flash Point 58°C (136°F)
Refractive Index 1.427
Quantity 50 g
UN Number UN3272
Beilstein 1721851
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material trans-2-Hexenyl acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.