missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TP53I13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79466
This item is not returnable.
View return policy
Description
TP53I13 Polyclonal specifically detects TP53I13 in Human samples. It is validated for Western Blot.
Specifications
| TP53I13 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DSCP1Damage-stimulated cytoplasmic protein 1, tumor protein p53 inducible protein 13, tumor protein p53-inducible protein 13 | |
| Rabbit | |
| 42 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Pig: 86%; Bovine: 86%; Horse: 85%; Rat: 79%; Mouse: 79%; Rabbit: 79%; Guinea pig: 79%. | |
| Human, Mouse, Rat, Pig, Bovine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_612358 | |
| TP53I13 | |
| Synthetic peptide directed towards the middle region of human TP53I13The immunogen for this antibody is TP53I13. Peptide sequence IYWGPTADSQDTVAAVLKRRLLQPSRRVKRSRRRPLLPPTPDSGPEGESS. | |
| Affinity purified | |
| RUO | |
| 90313 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction