Learn More
Tolnaftate, Thermo Scientific™
Antifungal agent
59.70€ - 568.00€
Chemical Identifiers
| CAS | 2398-96-1 |
|---|---|
| Molecular Formula | C19H17NOS |
| Molecular Weight (g/mol) | 307.411 |
| MDL Number | MFCD00056611 |
| InChI Key | FUSNMLFNXJSCDI-UHFFFAOYSA-N |
| Synonym | Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester; Carbamothioic acid |
| PubChem CID | 5510 |
| ChEBI | CHEBI:9620 |
| IUPAC Name | O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate |
| SMILES | CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15494329
|
Thermo Scientific Alfa Aesar
J61834.03 |
1g |
59.70€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15404339
|
Thermo Scientific Alfa Aesar
J61834.06 |
5g |
189.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15414339
|
Thermo Scientific Alfa Aesar
J61834.14 |
25g |
568.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
Chemical Identifiers
| 2398-96-1 | |
| 307.411 | |
| FUSNMLFNXJSCDI-UHFFFAOYSA-N | |
| 5510 | |
| O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate |
| C19H17NOS | |
| MFCD00056611 | |
| Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester; Carbamothioic acid | |
| CHEBI:9620 | |
| CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2 |
Specifications
| 2398-96-1 | |
| C19H17NOS | |
| 14,9519 | |
| Soluble in chloroform at 50 mg/ml; Sparingly soluble in ethanol or methanol | |
| CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2 | |
| 307.411 | |
| CHEBI:9620 | |
| Tolnaftate |
| White | |
| MFCD00056611 | |
| Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester; Carbamothioic acid | |
| FUSNMLFNXJSCDI-UHFFFAOYSA-N | |
| O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate | |
| 5510 | |
| 307.41 |
Safety and Handling
missing translation for 'einecsNumber' : 219-266-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.