Learn More
Tolnaftate, Thermo Scientific™
Antifungal agent
Brand: Thermo Scientific Alfa Aesar J61834.06
| Quantity | 5g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
Chemical Identifiers
| 2398-96-1 | |
| 307.411 | |
| FUSNMLFNXJSCDI-UHFFFAOYSA-N | |
| 5510 | |
| O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate |
| C19H17NOS | |
| MFCD00056611 | |
| Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester; Carbamothioic acid | |
| CHEBI:9620 | |
| CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2 |
Specifications
| 2398-96-1 | |
| 5g | |
| MFCD00056611 | |
| Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester; Carbamothioic acid | |
| FUSNMLFNXJSCDI-UHFFFAOYSA-N | |
| O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate | |
| 5510 | |
| 307.41 |
| White | |
| C19H17NOS | |
| 14,9519 | |
| Soluble in chloroform at 50 mg/ml; Sparingly soluble in ethanol or methanol | |
| CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2 | |
| 307.411 | |
| CHEBI:9620 | |
| Tolnaftate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.