accessibility menu, dialog, popup

UserName

CAS RN 2398-96-1

CH3NSOCH3

CAS RN 2398-96-1

IUPAC Name: N-methyl-N-(3-methylphenyl)-1-(naphthalen-2-yloxy)methanethioamide
Synonyms: tolnaftate tinactin jock itch cream genaspor aftate for athlete foot sprinkle powder tniaderm pitrex ony clear chinosol brand of tolnaftate tineafax tolnaftatum
Molecular Weight (g/mol): 307.41
Molecular Formula: C19H17NOS
InChi Key: FUSNMLFNXJSCDI-UHFFFAOYSA-N
SMILES: CN(C(=S)OC1=CC=C2C=CC=CC2=C1)C1=CC=CC(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Color 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)