Learn More
TNIK Mouse, anti-Human, CoraLite 594, Clone: 1E9H3, Proteintech
Mouse Monoclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | TNIK |
|---|---|
| Clone | 1E9H3 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite 594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18025395
|
Proteintech
CL594-67948-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18087275
|
Proteintech
CL594-67948-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TNIK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TNIK. This antibody recognizes human antigen. The TNIK antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™594.
Specifications
| TNIK | |
| 1000 μg/mL | |
| CoraLite 594 | |
| Mouse | |
| 23043 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1E9H3 | |
| Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human | |
| KIAA0551, TNIK | |
| Primary |