Learn More
TNIK Mouse, anti-Human, CoraLite 594, Clone: 1E9H3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-67948-20UL
 View more versions of this product
Product Code. 18025395
  • 145.00€ / 20µL
Description
The TNIK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TNIK. This antibody recognizes human antigen. The TNIK antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™594.
Specifications
Specifications
TNIK
1E9H3
CoraLite 594
Mouse
Primary
Human
155 kDa
IgG2a
Flow Cytometry (Intracellular Staining)
1000 μg/mL
KIAA0551, TNIK
20 μL
23043
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.