missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (TL2.1) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 38 publications
224.00€ - 560.00€
Specifications
| Antigen | TLR2 |
|---|---|
| Clone | TL2.1 |
| Dilution | Western Blot reported in scientific literature (PMID 28546218), Flow Cytometry 1ul/1 million cells, ELISA reported in scientific literature (PMID 23629653), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000. Use reported in scientific literature (Mempel et al (2003)), Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (Flo et al (2000)), Immunohistochemistry-Paraffin reported in scientific literature (PMID 11160251), In vitro assay reported in scientific literature (PMID 24512642), Flow (Cell Surface) 1-2 ug/1 million cells, Flow (Intracellular), CyTOF-ready, Block/Neutralize reported in scientific literature (Flo et al (2000); PMID 24512642) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084214
|
Novus Biologicals
NB100-56726SS |
0.025 mg |
224.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18791444
|
Novus Biologicals
NB100-56726 |
0.1 mg |
560.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR2 Monoclonal specifically detects TLR2 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready, Block/Neutralize.Specifications
| TLR2 | |
| Western Blot reported in scientific literature (PMID 28546218), Flow Cytometry 1ul/1 million cells, ELISA reported in scientific literature (PMID 23629653), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000. Use reported in scientific literature (Mempel et al (2003)), Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (Flo et al (2000)), Immunohistochemistry-Paraffin reported in scientific literature (PMID 11160251), In vitro assay reported in scientific literature (PMID 24512642), Flow (Cell Surface) 1-2 ug/1 million cells, Flow (Intracellular), CyTOF-ready, Block/Neutralize reported in scientific literature (Flo et al (2000); PMID 24512642) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Canine, Rabbit | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| TLR2 | |
| IgG2a | |
| Protein G purified |
| TL2.1 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity, Signal Transduction | |
| NP_003255.2 | |
| 7097 | |
| This antibody was raised by immunizing mice with CHO cells transfected with human TLR2 cDNA (Flo et al, 2000). The hybridoma supernatants were selected by flow cytometry. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title