missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody (TL2.1) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 38 publications
Brand: Novus Biologicals NB100-56726
This item is not returnable.
View return policy
Description
TLR2 Monoclonal specifically detects TLR2 in Human, Mouse, Rat, Canine, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, In vitro assay, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready, Block/Neutralize.
Specifications
| TLR2 | |
| Monoclonal | |
| 0.5mg/mL | |
| Western Blot reported in scientific literature (PMID 28546218), Flow Cytometry 1ul/1 million cells, ELISA reported in scientific literature (PMID 23629653), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000. Use reported in scientific literature (Mempel et al (2003)), Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (Flo et al (2000)), Immunohistochemistry-Paraffin reported in scientific literature (PMID 11160251), In vitro assay reported in scientific literature (PMID 24512642), Flow (Cell Surface) 1-2 ug/1 million cells, Flow (Intracellular), CyTOF-ready, Block/Neutralize reported in scientific literature (Flo et al (2000); PMID 24512642) | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody can be used in functional assays, such as inhibition of Listeria monocytogenes (HKLM) and LPS-induced IL-6 and TNF production. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| TL2.1 | |
| Unconjugated | |
| NP_003255.2 | |
| TLR2 | |
| This antibody was raised by immunizing mice with CHO cells transfected with human TLR2 cDNA (Flo et al, 2000). The hybridoma supernatants were selected by flow cytometry. | |
| 0.1 mg | |
| Immunology, Innate Immunity, Signal Transduction | |
| 7097 | |
| Human, Mouse, Rat, Canine, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction