missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 15 publications
240.00€ - 511.00€
Specifications
| Antigen | TLR2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry reported in scientific literature (PMID 31285077), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30618413), Immunoprecipitation reported in scientific literature (PMID 31383741), Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18707003
|
Novus Biologicals
NB100-56720SS |
0.025 mg |
240.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776463
|
Novus Biologicals
NB100-56720 |
0.1 mg |
511.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR2 Polyclonal specifically detects TLR2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TLR2 | |
| Western Blot 1-3 ug/ml, Flow Cytometry reported in scientific literature (PMID 31285077), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30618413), Immunoprecipitation reported in scientific literature (PMID 31383741), Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| O60603 | |
| 7097 | |
| This antibody was developed against a mixture of synthetic peptides containing amino acids 180-196, 353-370, and 473-489 of human TLR2 (NP_003255). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 89 kDa |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| PBS with 0.05% Sodium Azide | |
| CD282 antigen, TIL4CD282, toll/interleukin 1 receptor-like 4, Toll/interleukin-1 receptor-like protein 4, toll-like receptor 2 | |
| TLR2 | |
| IgG | |
| Protein G purified | |
| In transfected human TLR2 cell lysate, a 90 kDa band is observed. This has also been reported to work well in bronchial epithelial cells (UBH16E) and THP-1 cells. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel