Learn More
TLR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 15 publications
Brand: Novus Biologicals NB100-56720
This item is not returnable.
View return policy
Description
TLR2 Polyclonal specifically detects TLR2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| TLR2 | |
| 89 kDa | |
| RUO | |
| Purified |
| Western Blot 1-3 ug/ml, Flow Cytometry reported in scientific literature (PMID 31285077), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30618413), Immunoprecipitation reported in scientific literature (PMID 31383741), Immunohistochemistry-Paraffin 5 ug/ml | |
| Rabbit | |
| 0.1 mg | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |