Learn More
TLR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 15 publications
Brand:  Novus Biologicals NB100-56720
 View more versions of this product
Product Code. 18776463
  • 511.00€ / 0.10mg
Description
TLR2 Polyclonal specifically detects TLR2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
TLR2
89 kDa
RUO
Purified
Western Blot 1-3 ug/ml, Flow Cytometry reported in scientific literature (PMID 31285077), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 30618413), Immunoprecipitation reported in scientific literature (PMID 31383741), Immunohistochemistry-Paraffin 5 ug/ml
Rabbit
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.