missing translation for 'onlineSavingsMsg'
Learn More

TLE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen TLE3
Concentration 0.28 mg/mL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16801645
View Documents
Proteintech
22094-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16891635
View Documents
Proteintech
22094-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.
Specifications

Specifications

TLE3
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Rat, Mouse, Human
Q04726, Q08122, Q9JIT3
21887, 7090, 84424
TLE3 Fusion Protein Ag17300
Primary
-20°C
TLE3
0.28 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
ESG, ESG3, GRG3, HsT18976, KIAA1547, TLE3
TLE3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.