Learn More
TLE3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22094-1-AP-150UL
 View more versions of this product
Product Code. 16891635
  • 410.00€ / 150µL
Description
The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.
Specifications
Specifications
TLE3
Polyclonal
Unconjugated
TLE3
ESG, ESG3, GRG3, HsT18976, KIAA1547, TLE3
Rabbit
Antigen Affinity Chromatography
RUO
21887, 7090, 84424
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q04726, Q08122, Q9JIT3
TLE3
TLE3 Fusion Protein Ag17300
150 μL
Primary
Human, Mouse, Rat
Antibody