missing translation for 'onlineSavingsMsg'
Learn More

tescalcin Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

196.00€ - 468.00€

Specifications

Antigen tescalcin
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18632812
View Documents
Novus Biologicals
NBP2-94798-0.02ml
0.02 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
18646922
View Documents
Novus Biologicals
NBP2-94798-0.1ml
0.1 mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

tescalcin Polyclonal antibody specifically detects tescalcin in Human samples. It is validated for Western Blot
Specifications

Specifications

tescalcin
Western Blot
Unconjugated
Rabbit
Cancer, Cell Biology, Endocrinology, Neuroscience, Signal Transduction
PBS (pH 7.3), 50% glycerol
54997
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human
calcineurin B homologous protein 3, CHP3, TSC
Recombinant fusion protein containing a sequence corresponding to amino acids 135-214 of human TESC (NP_060369.3). YRNVVEELLSGNPHIEKESARSIADGAMMEAASVCMGQMEPDQVYEGITFEDFLKIWQGIDIETKMHVRFLNMETMALCH
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.