missing translation for 'onlineSavingsMsg'
Learn More

TCTE1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-94592-0.1ml

 View more versions of this product

Product Code. 18686891

  • 470.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TCTE1 Polyclonal antibody specifically detects TCTE1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCTE1
Polyclonal
Western Blot 1:500-1:2000
D6S46, FLJ50041, MGC33600, T-complex-associated testis-expressed protein 1, t-complex-associated-testis-expressed 1, tcte-1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TCTE1 (NP_872345.2). MQDTVTTSALLDPSHSSVSTQDNSSTGGHTSSTSPQLSKPSITPVPAKSRNPHPRANIRRMRRIIAEDPEWSLAIVPLLTELCIQHIIRNFQKNPILKQMLPEHQQKVLNHLSPDLPLAVTANLIDSENYWLRCCMHRWPVCHVAHHGGSWKRMFFERHLENLLKHFIPGTTDPAVILDLLPLCRNYVRRVHVDQFLPPVQLPAQLRPGDQSDSGSEGEMEEPTVDHYQLGDLVAGLSHLEELDLVYDVKDCGMNFEWNLFLFTYRDCLSLAAAIKACHTLKIFKLTRSKVDDDKARIII
0.1 mL
Cell Biology
202500
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.