missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 8b Antibody (MX-6), Novus Biologicals™
Mouse Monoclonal Antibody
195.00€
Specifications
| Antigen | TCR V beta 8b |
|---|---|
| Clone | MX-6 |
| Concentration | 0.15 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664809
|
Novus Biologicals
NBP2-22504-50ug |
50 μg | |||||||
Description
TCR V beta 8b Monoclonal specifically detects TCR V beta 8b in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| TCR V beta 8b | |
| 0.15 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
| Unconjugated | |
| Mouse | |
| Human | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| TCB | |
| IgG2a | |
| Protein G purified | |
| This antibody reacts with Human. |
| MX-6 | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
| Monoclonal | |
| Ascites | |
| RUO | |
| P04435 | |
| 6957 | |
| Human TCR Vbeta8 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title