missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 8b Antibody (MX-6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22504-50ug
This item is not returnable.
View return policy
Description
TCR V beta 8b Monoclonal specifically detects TCR V beta 8b in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| TCR V beta 8b | |
| Monoclonal | |
| 0.15 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6957 | |
| Human | |
| Ascites |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
| MX-6 | |
| Unconjugated | |
| P04435 | |
| TCB | |
| Human TCR Vbeta8 | |
| 50 μg | |
| Primary | |
| This antibody reacts with Human. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction