missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR gamma/delta Antibody (5A6.E9), Novus Biologicals™
Mouse Monoclonal Antibody
213.00€
Specifications
| Antigen | TCR gamma/delta |
|---|---|
| Clone | 5A6.E9 |
| Concentration | 0.15 mg/mL |
| Dilution | Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay |
Description
TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| TCR gamma/delta | |
| 0.15 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology, Stem Cells | |
| P09693 | |
| 6965 | |
| Human leukemic PEER cells | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 5A6.E9 | |
| Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG | |
| TRG | |
| IgG1 | |
| Protein A purified | |
| This antibody reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title