missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR gamma/delta Antibody (5A6.E9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22489-50ug
This item is not returnable.
View return policy
Description
TCR gamma/delta Monoclonal specifically detects TCR gamma/delta in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| TCR gamma/delta | |
| Monoclonal | |
| 0.15 mg/mL | |
| Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Functional | |
| T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody reacts with Human. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay | |
| 5A6.E9 | |
| Unconjugated | |
| P09693 | |
| TRG | |
| Human leukemic PEER cells | |
| 50 μg | |
| Adaptive Immunity, Immunology, Stem Cells | |
| 6965 | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction