missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 alpha Antibody (91a), Novus Biologicals™
Rat Monoclonal Antibody
193.00€ - 681.00€
Specifications
| Antigen | TCP1 alpha |
|---|---|
| Clone | 91a |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066627
|
Novus Biologicals
NBP1-97505-0.025MG |
0.025 mg |
193.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18047238
|
Novus Biologicals
NBP1-97505-0.05MG |
0.05 mg |
418.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18087418
|
Novus Biologicals
NBP1-97505-0.2MG |
0.2 mg |
681.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCP1 alpha Monoclonal specifically detects TCP1 alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Drosophila, Guinea Pig, Plant, Primate, Rabbit, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| TCP1 alpha | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Yeast, Mouse, Guinea Pig, Rabbit, Primate, Human, Canine, Pig, C. elegans, Rat, Bovine, Drosophila, Plant | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| TCP1 | |
| IgG2a | |
| Protein G purified |
| 91a | |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rat | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| 6950 | |
| Recombinant mouse TCP-1alpha (C-terminal fragment). | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title