missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 alpha Antibody (91a), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-97505-0.05MG
This item is not returnable.
View return policy
Description
TCP1 alpha Monoclonal specifically detects TCP1 alpha in Human, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Drosophila, Guinea Pig, Plant, Primate, Rabbit, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| TCP1 alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| TCP1 | |
| Recombinant mouse TCP-1alpha (C-terminal fragment). | |
| 0.05 mg | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 6950 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 91a | |
| Western Blot 1:1000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Canine, C. elegans, Drosophila, Guinea Pig, Plant, Primate, Rabbit, Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction