missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 alpha Antibody (23c (CTA-123)) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
190.00€ - 351.00€
Specifications
| Antigen | TCP1 alpha |
|---|---|
| Clone | 23c (CTA-123) |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626837
|
Novus Biologicals
NBP2-50328-0.025ml |
0.025 mL |
190.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626636
|
Novus Biologicals
NBP2-50328 |
0.1 mL |
351.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCP1 alpha Monoclonal antibody specifically detects TCP1 alpha in Mouse, Rat, Canine, Chicken, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| TCP1 alpha | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| PBS | |
| 6950 | |
| IgG2c | |
| Protein G purified |
| 23c (CTA-123) | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat, Canine, Chicken, Hamster | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| C-terminal half of the full length murine TCP protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title