missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP1 alpha Antibody (23c (CTA-123)) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50328-0.025ml
This item is not returnable.
View return policy
Description
TCP1 alpha Monoclonal antibody specifically detects TCP1 alpha in Mouse, Rat, Canine, Chicken, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| TCP1 alpha | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| CCT1T-complex protein 1 subunit alpha, CCTa, CCT-alpha, D6S230E, tailless complex polypeptide 1, t-complex 1, T-complex protein 1, alpha subunit, TCP-1-alpha | |
| C-terminal half of the full length murine TCP protein | |
| 0.025 mL | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 6950 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2c |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| 23c (CTA-123) | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat, Canine, Chicken, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction