missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-3/E2A Antibody (6B8), Novus Biologicals™
Mouse Monoclonal Antibody
217.00€
Specifications
| Antigen | TCF-3/E2A |
|---|---|
| Clone | 6B8 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
TCF-3/E2A Monoclonal specifically detects TCF-3/E2A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| TCF-3/E2A | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| BHLHB21, bHLHb21TCF-3, Class B basic helix-loop-helix protein 21, E2A immunoglobulin enhancer-binding factor E12/E47, E2AKappa-E2-binding factor, Immunoglobulin enhancer-binding factor E12/E47, Immunoglobulin transcription factor 1, ITF1Transcription factor ITF-1, MGC129647, MGC129648, Transcription factor 3, transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47), transcription factor E2-alpha, VDIR, VDR interacting repressor | |
| TCF3 | |
| IgG1 | |
| Unpurified | |
| 67.6 kDa |
| 6B8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Transcription Factors and Regulators | |
| Ascites with 0.03% Sodium Azide | |
| 6929 | |
| Purified recombinant fragment of human TCF3 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title