missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF-3/E2A Antibody (6B8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-42566-0.025ml
This item is not returnable.
View return policy
Description
TCF-3/E2A Monoclonal specifically detects TCF-3/E2A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| TCF-3/E2A | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| TCF3 | |
| Purified recombinant fragment of human TCF3 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot | |
| 6B8 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| BHLHB21, bHLHb21TCF-3, Class B basic helix-loop-helix protein 21, E2A immunoglobulin enhancer-binding factor E12/E47, E2AKappa-E2-binding factor, Immunoglobulin enhancer-binding factor E12/E47, Immunoglobulin transcription factor 1, ITF1Transcription factor ITF-1, MGC129647, MGC129648, Transcription factor 3, transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47), transcription factor E2-alpha, VDIR, VDR interacting repressor | |
| Mouse | |
| 67.6 kDa | |
| 0.025 mL | |
| Transcription Factors and Regulators | |
| 6929 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction