missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACE/ADAM17 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
189.00€
Specifications
| Antigen | TACE/ADAM17 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.25-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
TACE/ADAM17 Polyclonal specifically detects TACE/ADAM17 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| TACE/ADAM17 | |
| Western Blot 0.25-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Stem Cell Signaling Pathway | |
| PBS with 0.02% Sodium Azide | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM18, CD156B, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| ADAM17 | |
| IgG | |
| Affinity Purified | |
| 80 to 130 kDa bands can be detected, which may represent mature protein, precursor, and glycosylated TACE/ADAM17. Human TACE/ADAM17 has 2 isoforms, including isoform A (824aa, 93kD), isoform B (694aa, 78.5kD). This antibody detects only isoform A. Mouse TACE/ADAM17 has 2 isoforms, long isoform (827aa, 93.1kD) and short isoform (655aa, 73.9kD). Rat TACE/ADAM17 has only one isoform identified so far (827aa, 93kD). |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Rat | |
| NP_003174 | |
| 6868 | |
| Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TACE. The immunogen is located within the last 50 amino acids of TACE. Amino Acid Squence: ASFKLQRQNRVDSKETE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| 93 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title