missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACE/ADAM17 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-77044-0.025mg
This item is not returnable.
View return policy
Description
TACE/ADAM17 Polyclonal specifically detects TACE/ADAM17 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| TACE/ADAM17 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM18, CD156B, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| Rabbit | |
| 93 kDa | |
| 0.025 mg | |
| Apoptosis, Stem Cell Signaling Pathway | |
| 6868 | |
| Human, Rat | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.25-2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10 μg/mL | |
| NP_003174 | |
| ADAM17 | |
| Antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TACE. The immunogen is located within the last 50 amino acids of TACE. Amino Acid Squence: ASFKLQRQNRVDSKETE | |
| Affinity Purified | |
| RUO | |
| Primary | |
| 80 to 130 kDa bands can be detected, which may represent mature protein, precursor, and glycosylated TACE/ADAM17. Human TACE/ADAM17 has 2 isoforms, including isoform A (824aa, 93kD), isoform B (694aa, 78.5kD). This antibody detects only isoform A. Mouse TACE/ADAM17 has 2 isoforms, long isoform (827aa, 93.1kD) and short isoform (655aa, 73.9kD). Rat TACE/ADAM17 has only one isoform identified so far (827aa, 93kD). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction