missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T7 RNA polymerase Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | T7 RNA polymerase |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234414
|
Proteintech
29943-1-AP-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237852
|
Proteintech
29943-1-AP-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The T7 RNA polymerase antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of bacteriophage t7 T7 RNA polymerase. This antibody recognizes bacteriophage t7 antigen. The T7 RNA polymerase antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| T7 RNA polymerase | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Bacteriophage T7 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 600 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 1261050 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title