missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T7 RNA polymerase Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29943-1-AP-150UL
This item is not returnable.
View return policy
Description
The T7 RNA polymerase antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of bacteriophage t7 T7 RNA polymerase. This antibody recognizes bacteriophage t7 antigen. The T7 RNA polymerase antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| T7 RNA polymerase | |
| Polyclonal | |
| Unconjugated | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Bacteriophage T7 | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 600 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1261050 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction