missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synapsin I Antibody (7B1D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€
Specifications
| Antigen | Synapsin I |
|---|---|
| Clone | 7B1D9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Classification | Monoclonal |
Description
Synapsin I Monoclonal specifically detects Synapsin I in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Synapsin I | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Cellular Markers, Circadian Rhythm, Cognition and Behavior, Neuronal Cell Markers, Neuroscience, Neurotransmission, Pain Research, Phospho Specific, Sensory Systems, Vision | |
| SYN1a, SYN1b, Synapsin 1, synapsin IBrain protein 4.1, synapsin-1, SYNI | |
| SYN1 | |
| IgG1 | |
| Protein G purified |
| 7B1D9 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 6853 | |
| Purified recombinant fragment of human SYN1 (AA: 362-511) expressed in E. Coli. | |
| Primary | |
| 74.1 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title