missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synapsin I Antibody (7B1D9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61894-0.025ml
This item is not returnable.
View return policy
Description
Synapsin I Monoclonal specifically detects Synapsin I in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Synapsin I | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| SYN1a, SYN1b, Synapsin 1, synapsin IBrain protein 4.1, synapsin-1, SYNI | |
| Mouse | |
| 74.1 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 7B1D9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SYN1 | |
| Purified recombinant fragment of human SYN1 (AA: 362-511) expressed in E. Coli. | |
| Protein G purified | |
| Cellular Markers, Circadian Rhythm, Cognition and Behavior, Neuronal Cell Markers, Neuroscience, Neurotransmission, Pain Research, Phospho Specific, Sensory Systems, Vision | |
| 6853 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction