missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYK Antibody (2G11E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
217.00€
Specifications
| Antigen | SYK |
|---|---|
| Clone | 2G11E6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Description
SYK Monoclonal specifically detects SYK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| SYK | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| DKFZp313N1010, EC 2.7.10, EC 2.7.10.2, FLJ25043, spleen tyrosine kinaseFLJ37489, tyrosine-protein kinase SYK | |
| SYK | |
| IgG1 | |
| Protein G purified |
| 2G11E6 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
| Monoclonal | |
| Purified | |
| Immunology, Innate Immunity, Mast Cell Markers, Phospho Specific, Protein Kinase | |
| 6850 | |
| Purified recombinant fragment of human SYK (AA: 217-356) expressed in E. Coli. | |
| Primary | |
| 57.2 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title