missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYK Antibody (2G11E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61893-0.025ml
This item is not returnable.
View return policy
Description
SYK Monoclonal specifically detects SYK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| SYK | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
| DKFZp313N1010, EC 2.7.10, EC 2.7.10.2, FLJ25043, spleen tyrosine kinaseFLJ37489, tyrosine-protein kinase SYK | |
| Mouse | |
| 57.2 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 2G11E6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SYK | |
| Purified recombinant fragment of human SYK (AA: 217-356) expressed in E. Coli. | |
| Protein G purified | |
| Immunology, Innate Immunity, Mast Cell Markers, Phospho Specific, Protein Kinase | |
| 6850 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction