missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV40 T-antigen Antibody (Pab 430) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
214.00€ - 430.00€
Specifications
| Antigen | SV40 T-antigen |
|---|---|
| Clone | Pab 430 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 |
| Applications | Western Blot, Immunoprecipitation, Radioimmune Assays (RIA) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18135918
|
Novus Biologicals
NBP2-50598 |
0.1 mL |
430.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665316
|
Novus Biologicals
NBP2-50598-0.025ml |
0.025 mL |
214.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SV40 T-antigen Monoclonal specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, Immunoprecipitation, Radioimmunoassay.Specifications
| SV40 T-antigen | |
| 1.0 mg/mL | |
| Western Blot, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Mouse | |
| Cancer, Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| SV40 transformed mouse cells | |
| Primary | |
| Specific for Simian virus (SV40) T antigen (large T and small T antigen). |
| Pab 430 | |
| Western Blot 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Large T antigen, Small T antigen | |
| IgG3 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title