Learn More
SV40 T-antigen Antibody (Pab 430) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-50598
This item is not returnable.
View return policy
Description
SV40 T-antigen Monoclonal specifically detects SV40 T-antigen in Virus samples. It is validated for Western Blot, Immunoprecipitation, Radioimmunoassay.
Specifications
| SV40 T-antigen | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| Large T antigen, Small T antigen | |
| SV40 transformed mouse cells | |
| 0.1 mL | |
| Cancer, Virology Bacteria and Parasites | |
| Specific for Simian virus (SV40) T antigen (large T and small T antigen). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG3 |
| Western Blot, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Pab 430 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |