missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV2B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 470.00€
Specifications
| Antigen | SV2B |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637212
|
Novus Biologicals
NBP2-94221-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611272
|
Novus Biologicals
NBP2-94221-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SV2B Polyclonal antibody specifically detects SV2B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SV2B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 9899 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ30413, FLJ42732, KIAA0735HsT19680, synaptic vesicle glycoprotein 2B, synaptic vesicle protein 2B homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 415-540 of human SV2B (NP_055663.1). IRYFQDEEYKSKMKVFFGEHVYGATINFTMENQIHQHGKLVNDKFTRMYFKHVLFEDTFFDECYFEDVTSTDTYFKNCTIESTIFYNTDLYEHKFINCRFINSTFLEQKEGCHMDLEQDNDFLIYL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title