missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV2B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94221-0.02ml
This item is not returnable.
View return policy
Description
SV2B Polyclonal antibody specifically detects SV2B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SV2B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ30413, FLJ42732, KIAA0735HsT19680, synaptic vesicle glycoprotein 2B, synaptic vesicle protein 2B homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 415-540 of human SV2B (NP_055663.1). IRYFQDEEYKSKMKVFFGEHVYGATINFTMENQIHQHGKLVNDKFTRMYFKHVLFEDTFFDECYFEDVTSTDTYFKNCTIESTIFYNTDLYEHKFINCRFINSTFLEQKEGCHMDLEQDNDFLIYL | |
| 0.02 mL | |
| Neuroscience | |
| 9899 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction