missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SURF4 Antibody (S07-1F1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19808-100ul
This item is not returnable.
View return policy
Description
SURF4 Monoclonal antibody specifically detects SURF4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SURF4 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6836 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| S07-1F1 | |
| Unconjugated | |
| ERV29, FLJ22993, MGC102753, SURF-4, surface 4 integral membrane protein, surfeit 4, surfeit locus protein 4 | |
| A synthetic peptide of human SURF4 (Uniprot # O15260) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction