missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SURF4 Antibody (S07-1F1), Novus Biologicals™
Rabbit Monoclonal Antibody
444.00€ - 680.00€
Specifications
| Antigen | SURF4 |
|---|---|
| Clone | S07-1F1 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607603
|
Novus Biologicals
NBP3-19808-100ul |
100 μg |
680.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655753
|
Novus Biologicals
NBP3-19808-50ul |
50 μg |
444.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SURF4 Monoclonal antibody specifically detects SURF4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SURF4 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 6836 | |
| IgG | |
| Affinity purified |
| S07-1F1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ERV29, FLJ22993, MGC102753, SURF-4, surface 4 integral membrane protein, surfeit 4, surfeit locus protein 4 | |
| A synthetic peptide of human SURF4 (Uniprot # O15260) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title