missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | STRN3 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16822565
|
Proteintech
23966-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16812565
|
Proteintech
23966-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SG2NA was identified as a human cancer autoantigen and has now been implicated in vesicular trafficking and signal transduction. It is highly related to striatin and zinedin. It is a scaffolding protein that binds PP2A A and C subunits as well as the human homolog of the yeast protein, Mob1, which has been implicated in control of mitotic progression in yeast. It also binds calmodulin and appears to modulate PP2A activity.Specifications
| STRN3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P58405, Q13033, Q9ERG2 | |
| 114520, 29966, 94186 | |
| STRN3 Fusion Protein Ag21091 | |
| Primary | |
| -20°C | |
| STRN3 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Cell cycle autoantigen SG2NA, GS2NA, S/G2 antigen, SG2NA, Striatin 3, STRN3 | |
| Strn3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title