missing translation for 'onlineSavingsMsg'
Learn More

STRN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16822565 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16822565 20 μL 20µL
16812565 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16822565 Supplier Proteintech Supplier No. 239661AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SG2NA was identified as a human cancer autoantigen and has now been implicated in vesicular trafficking and signal transduction. It is highly related to striatin and zinedin. It is a scaffolding protein that binds PP2A A and C subunits as well as the human homolog of the yeast protein, Mob1, which has been implicated in control of mitotic progression in yeast. It also binds calmodulin and appears to modulate PP2A activity.

Specifications

Antigen STRN3
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STRN3
Gene Accession No. P58405, Q13033, Q9ERG2
Gene Alias Cell cycle autoantigen SG2NA, GS2NA, S/G2 antigen, SG2NA, Striatin 3, STRN3
Gene Symbols Strn3
Host Species Rabbit
Immunogen STRN3 Fusion Protein Ag21091
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114520, 29966, 94186
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.