missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stomatin Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | Stomatin |
|---|---|
| Clone | 1A4D6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234561
|
Proteintech
67894-1-IG-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233336
|
Proteintech
67894-1-IG-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Stomatin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Stomatin. This antibody recognizes human antigen. The Stomatin antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| Stomatin | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 2040 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. |
| 1A4D6 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| BND7; EPB7; EPB72 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title