missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3 Antibody (9D8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
215.00€
Specifications
| Antigen | STAT3 |
|---|---|
| Clone | 9D8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000, Immunohistochemistry 1:1600, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:1600, Chromatin Immunoprecipitation (ChIP) 1-3 ul, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Description
STAT3 Monoclonal specifically detects STAT3 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| STAT3 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Signal Transduction, Transcription Factors and Regulators | |
| P40763 | |
| 6774 | |
| Residues 655-770 from human recombinant protein expressed in bacteria | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9D8 | |
| Western Blot 1:5000, Immunohistochemistry 1:1600, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:1600, Chromatin Immunoprecipitation (ChIP) 1-3 ul, Knockdown Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Primate, Monkey | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| STAT3 | |
| IgG2a | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title