Learn More
STAT3 Antibody (9D8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP2-22471-0.025ml
 View more versions of this product
Product Code. 18763114
  • ONLINE EXCLUSIVE PRICE
    215.00€
    202.65€ / 0.03mL
    Save 12.35€
    5% Off
Description
STAT3 Monoclonal specifically detects STAT3 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT3
Monoclonal
1 mg/mL
Western Blot 1:5000, Immunohistochemistry 1:1600, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:1600, Chromatin Immunoprecipitation (ChIP) 1-3 ul, Knockdown Validated
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat, Primate, Monkey
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
9D8
Unconjugated
P40763
STAT3
Residues 655-770 from human recombinant protein expressed in bacteria
0.025 mL
Cellular Markers, Signal Transduction, Transcription Factors and Regulators
6774
Store at -20°C. Avoid freeze-thaw cycles.
IgG2a