Learn More
Standard Buffer Solution pH 4.01, WTW™
GREENER_CHOICE
Product Code. 11705214
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11705214 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11705214 Supplier WTW Supplier No. 109110
  • ONLINE EXCLUSIVE PRICE
    34.17€
    31.20€ / 250mL
    Save 2.97€
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
With a high accuracy: ±0.02 pH, this standard buffering solution is traceably certified to PTB/NIST and therefore safe for use.
  • For precision applications
  • PTB/NIST traceable standard buffer solutions (work reference buffer solutions) for precision applications - safe use
  • Universal buffering solutions
  • High accuracy: ±0.02 pH
  • Buffering solutions as per DIN 19266-01: 2000 are the most widely distributed buffering solutions
  • Standard buffering solutions by WTW are traceably certified to PTB/NIST and therefore safe for use
  • As standard buffering solutions, the buffer pH 4.006 and PL 4 are available in 250 ml bottles
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 100°C
Color Colorless
Certificate of Analysis DIN/NBS
Density 1.13g/cm3
For Use With (Application) Precision applications
pH 4.01
Physical Form Liquid
Quantity 250 mL
Packaging Bottle