missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
218.00€
Specifications
| Antigen | SREBP1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:5000, Immunocytochemistry/Immunofluorescence 1:50 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
SREBP1 Polyclonal specifically detects SREBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SREBP1 | |
| Western Blot 1:500 - 1:5000, Immunocytochemistry/Immunofluorescence 1:50 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
| BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
| SREBF1 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9WTN3 | |
| 6720 | |
| Synthetic peptide corresponding to residues M(32) L Q L I N N Q D S D F P G L F(47) of mouse SREBP 1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title