missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SREBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-74542-0.025mg
This item is not returnable.
View return policy
Description
SREBP1 Polyclonal specifically detects SREBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SREBP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
| BHLHD1, Class D basic helix-loop-helix protein 1, SREBP 1c, SREBP-1, SREBP1bHLHd1SREBP-1c, sterol regulatory element binding protein-1, sterol regulatory element binding transcription factor 1, sterol regulatory element-binding protein 1, Sterol regulatory element-binding transcription factor 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:5000, Immunocytochemistry/Immunofluorescence 1:50 - 1:500 | |
| Q9WTN3 | |
| SREBF1 | |
| Synthetic peptide corresponding to residues M(32) L Q L I N N Q D S D F P G L F(47) of mouse SREBP 1. | |
| 0.025 mg | |
| Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators | |
| 6720 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction