missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin beta 1 Antibody (4C3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
213.00€
Specifications
| Antigen | Spectrin beta 1 |
|---|---|
| Clone | 4C3 |
| Dilution | Western Blot 1:100, Flow Cytometry 0.5 - 2 μg, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Spectrin beta 1 Monoclonal specifically detects Spectrin beta 1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Spectrin beta 1 | |
| Western Blot 1:100, Flow Cytometry 0.5 - 2 μg, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunohistochemistry-Paraffin 1:20 - 1:200, Immunohistochemistry-Frozen 1:20 - 1:200 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 6710 | |
| Purified human erythrocyte beta-1 spectrin. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 4C3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cytoskeleton Markers | |
| beta-I spectrin, EL3, erythrocyte, HS2, HSPTB1, membrane cytoskeletal protein, spectrin, beta, erythrocytic, SPTB1 | |
| SPTB | |
| IgG1 | |
| Unpurified | |
| Detects spectrin from erythrocytes, brain and muscle cells. This has been shown to specifically detect the two known alternatively spliced forms of spectrin, beta-1 epsilon-1, present in erythrocytes, and beta-1 epsilon-2, present in nerve and striated muscle cells. It does not cross-react with alpha-2 spectrin or either of the fodrin subunits. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title